missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromothiophene-2-carboxylate 98.0+%, TCI America™

Catalog No. M25901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25901G 1 g
M25905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25901G Supplier TCI America Supplier No. M25901G

Chemical Identifiers

CAS 26137-08-6
Molecular Formula C6H5BrO2S
Molecular Weight (g/mol) 221.068
MDL Number MFCD00173839
InChI Key PEGSJNCGPSIJOX-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxylic acid methyl ester, 3-bromo-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-bromo-, methyl ester, 3-bromo-2-thiophenecarboxylic acid methyl ester, methyl 3-bromo-2-thiophenecarboxylate, maybridge3_006386, ksc494q2p, 2-methoxycarbonyl-3-bromothiophene, methyl-3-bromothiophene carboxylate, methyl 3-bromanylthiophene-2-carboxylate
PubChem CID 2740074
IUPAC Name methyl 3-bromothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)Br

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 221.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Bromothiophene-2-carboxylate