missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromopropionate 99.0+%, TCI America™

Catalog No. p-7118589
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B064825G 25 g
B0648100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B064825G Supplier TCI America Supplier No. B064825G
Only null left

Chemical Identifiers

CAS 3395-91-3
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000250
InChI Key KQEVIFKPZOGBMZ-UHFFFAOYSA-N
Synonym methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
PubChem CID 76934
IUPAC Name methyl 3-bromopropanoate
SMILES COC(=O)CCBr

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 g
Formula Weight 167.00
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Bromopropionate
RTECSNumber UA2451790
TSCA Yes