missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromopropionate, 97%

Catalog No. AAL1412714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1412714 25 g
AAL1412722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1412714 Supplier Thermo Scientific Chemicals Supplier No. L1412714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3395-91-3 | C4H7BrO2 | 167.00 g/mol

Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 3395-91-3
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000250
InChI Key KQEVIFKPZOGBMZ-UHFFFAOYSA-N
Synonym methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
PubChem CID 76934
IUPAC Name methyl 3-bromopropanoate
SMILES COC(=O)CCBr

Specifications

Density 1.53
Boiling Point 58°C to 60°C (12 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.458
Quantity 25 g
Beilstein 1742949
Solubility Information Immiscible in water.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material Methyl 3-bromopropionate
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 222-247-5
RTECSNumber UA2451790
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only