missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromopropionate, 97%

Catalog No. p-161009
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221791000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC221791000 Supplier Thermo Scientific Chemicals Supplier No. 221791000
Only null left

CAS: 3395-91-3 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3395-91-3
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000250
InChI Key KQEVIFKPZOGBMZ-UHFFFAOYSA-N
Synonym methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
PubChem CID 76934
IUPAC Name methyl 3-bromopropanoate
SMILES COC(=O)CCBr

Specifications

Color Colorless to Yellow
Density 1.5300g/mL
Boiling Point 64°C to 66°C (18.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4565 to 1.4595
Linear Formula BrCH2CH2CO2CH3
Quantity 100 mL
Beilstein 02, II, 231
Specific Gravity 1.53
Merck Index 15, 1444
Solubility Information Solubility in water: immiscible
Formula Weight 167
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 3-bromopropionate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-247-5
RTECSNumber UA2451790
TSCA TSCA

RUO – Research Use Only