missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH3599403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3599403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3599403 Supplier Thermo Scientific Chemicals Supplier No. H3599403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 342613-63-2 | C9H7BrN2O2 | 255.071 g/mol

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 342613-63-2
Molecular Formula C9H7BrN2O2
Molecular Weight (g/mol) 255.071
MDL Number MFCD10566741
InChI Key VFFFBWRUZMAHFS-UHFFFAOYSA-N
Synonym methyl 3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-7-methoxycarbonyl imidazo 1,2-a pyridine, methyl 3-bromoh-imidazo 1,2-a pyridine-7-carboxylate, 3-bromoimidazo 1,2-a pyridine-7-carboxylic acid methyl ester, pubchem23890, methyl3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, methyl 3-bromo-5h-imidazo 2,1-b 1,3 oxazine-7-carboxylate, 3-bromo-imidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester, 3-bromoimidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester
PubChem CID 45790121
IUPAC Name methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
SMILES COC(=O)C1=CC2=NC=C(N2C=C1)Br

Specifications

Melting Point 156°C to 157°C
Quantity 1 g
Formula Weight 255.07
Percent Purity 95%
Chemical Name or Material Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only