missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-2-fluorophenylacetate, 96%, Thermo Scientific™

Catalog No. p-7054446
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6265503 1g
AAH6265506 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6265503 Supplier Thermo Scientific Chemicals Supplier No. H6265503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1427451-43-1
Molecular Formula C9H8BrFO2
Molecular Weight (g/mol) 247.063
InChI Key ALPXGXOFMZOFAG-UHFFFAOYSA-N
Synonym methyl 3-bromo-2-fluorophenylacetate, methyl 2-3-bromo-2-fluorophenyl acetate
PubChem CID 100912177
IUPAC Name methyl 2-(3-bromo-2-fluorophenyl)acetate
SMILES COC(=O)CC1=C(C(=CC=C1)Br)F

Specifications

Quantity 1g
Formula Weight 247.06
Percent Purity 96%
Chemical Name or Material Methyl 3-Bromo-2-fluorophenylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only