missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-2-(bromomethyl)propionate 97.0+%, TCI America™

Supplier:  TCI America M206625G

 View more versions of this product

Catalog No. M206625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl 3-Bromo-2-(bromomethyl)propionate
62°C
MFCD00011534
methyl 3-bromo-2-bromomethyl propionate, methyl 3-bromo-2-bromomethyl propanoate, methyl3-bromo-2-bromomethyl propionate, methyl bis bromomethyl acetate, methyl 2-bis-bromomethyl-acetate, propanoic acid, 3-bromo-2-bromomethyl-, methyl ester, propanoic acid,3-bromo-2-bromomethyl-, methyl ester, zlchem 71, acmc-1chw2, 3-bromo-2-bromomethyl-propionic acid methyl ester
COC(=O)C(CBr)CBr
259.925
259.93
Liquid
22262-60-8
C5H8Br2O2
25 g
USXVPPOARMSYGY-UHFFFAOYSA-N
methyl 3-bromo-2-(bromomethyl)propanoate
140895
≥97.0% (GC)

Chemical Identifiers

22262-60-8
259.925
USXVPPOARMSYGY-UHFFFAOYSA-N
methyl 3-bromo-2-(bromomethyl)propanoate
C5H8Br2O2
MFCD00011534
140895
COC(=O)C(CBr)CBr
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions