missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Benzoylpropionate 98.0+%, TCI America™

Catalog No. p-7119110
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15465G 5 g
B154625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15465G Supplier TCI America Supplier No. B15465G

Chemical Identifiers

CAS 25333-24-8
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00014906
InChI Key XVRCVKWYKYJEIG-UHFFFAOYSA-N
Synonym methyl 3-benzoylpropionate, 3-benzoylpropionic acid methyl ester, propionic acid, 3-benzoyl-, methyl ester, methyl 4-oxo-4-phenylbutyrate, benzenebutanoic acid, gamma-oxo-, methyl ester, benzenebutanoic acid, .gamma.-oxo-, methyl ester, benzenebutanoic acid, g-oxo-, methyl ester, acmc-209gja, methyl3-benzoylpropionate, methyl-3-benzoylpropionate
PubChem CID 141190
IUPAC Name methyl 4-oxo-4-phenylbutanoate
SMILES COC(=O)CCC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Benzoylpropionate