missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminothiophene-2-carboxylate, 98+%

Catalog No. AAA1280222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1280222 100 g
AAA1280206 5 g
AAA1280214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1280222 Supplier Thermo Scientific Chemicals Supplier No. A1280222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22288-78-4 | C6H7NO2S | 157.19 g/mol

Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 22288-78-4
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00009765
InChI Key TWEQNZZOOFKOER-UHFFFAOYSA-N
Synonym methyl 3-amino-2-thiophenecarboxylate, 3-aminothiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-, methyl ester, 3-amino-thiophene-2-carboxylic acid methyl ester, 3-amino-2-thiophenecarboxylic acid methyl ester, methyl 3-amino-2-thiophene carboxylate, methyl-3-amino-thiophene-2-carboxylate, 3-amino-2-carbomethoxythiophene, 3-amino-2-methoxycarbonylthiophene, methyl3-aminothiophene-2-carboxylate
PubChem CID 89652
IUPAC Name methyl 3-aminothiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C=CS1

Specifications

Melting Point 63°C to 67°C
Boiling Point 100°C to 102°C (0.1 mmHg)
Quantity 100 g
Beilstein 1365614
Solubility Information Slightly soluble in water.
Formula Weight 157.19
Percent Purity ≥98%
Chemical Name or Material Methyl 3-aminothiophene-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-895-8
RTECSNumber XM8347500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only