missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Aminopyrazine-2-carboxylate 98.0+%, TCI America™

Catalog No. A11865G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11865G 5 g
A11861G 1 g
A118625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A11865G Supplier TCI America Supplier No. A11865G
Only null left

Chemical Identifiers

CAS 16298-03-6
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00010097
InChI Key INCSQLZZXBPATR-UHFFFAOYSA-N
Synonym methyl 3-amino-2-pyrazinecarboxylate, methyl 2-aminopyrazine-3-carboxylate, 3-aminopyrazine-2-carboxylic acid methyl ester, methyl 3-aminopyrazinecarboxylate, methyl 3-amino-2-pyrazine carboxylate, 3-amino-pyrazine-2-carboxylic acid methyl ester, 3-amino-2-pyrazinecarboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-, methyl ester, methyl-3-aminopyrazin-2-carboxylat, zlchem 76
PubChem CID 72669
IUPAC Name methyl 3-aminopyrazine-2-carboxylate
SMILES COC(=O)C1=NC=CN=C1N

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Aminopyrazine-2-carboxylate