missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminocrotonate, 97%

Catalog No. AC156790050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156790050 5 g Glass bottle
AC156791000 100 g Plastic bottle
AC156795000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156790050 Supplier Thermo Scientific Chemicals Supplier No. 156790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14205-39-1 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14205-39-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00008072,MFCD00008072,MFCD00008072
InChI Key XKORCTIIRYKLLG-ONEGZZNKSA-N
Synonym methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester
PubChem CID 643918
IUPAC Name methyl (Z)-3-aminobut-2-enoate
SMILES COC(=O)\C=C(/C)N

Specifications

Melting Point 81.0°C to 83.0°C
Color White to Yellow
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C(NH2)=CHCO2CH3
Quantity 5 g
Beilstein 03, 632
Solubility Information Solubility in water: insoluble in water. Other solubilities: solubility in chloroform >500g/L (20°C),solubility in acetone >500g/L (20°C)
Formula Weight 115.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-aminocrotonate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-056-5
RTECSNumber EM9092500
TSCA TSCA

RUO – Research Use Only