missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminobenzoate, 98%

Catalog No. AC413950050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413950050 5 g Glass bottle
AC413950250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413950050 Supplier Thermo Scientific Chemicals Supplier No. 413950050

CAS: 4518-10-9 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4518-10-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00017102
InChI Key VZDNXXPBYLGWOS-UHFFFAOYSA-N

Specifications

Melting Point 52°C to 55°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Synonym benzoic acid, 3-amino-, methyl ester, methyl-3-aminobenzoate, 3-aminobenzoic acid methyl ester, m-aminobenzoic acid, methyl ester, 3-amino-benzoic acid methyl ester, methyl m-aminobenzoate, methyl3-aminobenzoate, methyl-m-aminobenzoate, pubchem16709, acmc-1aeif
SMILES COC(=O)C1=CC(=CC=C1)N
IUPAC Name methyl 3-aminobenzoate
PubChem CID 78274
Formula Weight 151.16
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 3-aminobenzoate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-842-5
TSCA TSCA

RUO – Research Use Only