missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminobenzoate, 98%

Catalog No. p-7022136
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1476606 5 g
AAA1476614 25 g
AAA1476622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1476606 Supplier Thermo Scientific Chemicals Supplier No. A1476606
Only null left

CAS: 4518-10-9 | C8H9NO2 | 151.17 g/mol

Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water (0.1-1%) .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 4518-10-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017102
InChI Key VZDNXXPBYLGWOS-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-, methyl ester, methyl-3-aminobenzoate, 3-aminobenzoic acid methyl ester, m-aminobenzoic acid, methyl ester, 3-amino-benzoic acid methyl ester, methyl m-aminobenzoate, methyl3-aminobenzoate, methyl-m-aminobenzoate, pubchem16709, acmc-1aeif
PubChem CID 78274
IUPAC Name methyl 3-aminobenzoate
SMILES COC(=O)C1=CC(=CC=C1)N

Specifications

Melting Point 49°C to 53°C
Density 1.232
Quantity 5 g
Beilstein 1101468
Solubility Information Slightly soluble in water (0.1-1%) .
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material Methyl 3-aminobenzoate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-842-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only