missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 35212-85-2
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.25
MDL Number MFCD00206744
InChI Key VLHHEYMZLXKSQO-UHFFFAOYSA-N
Synonym methyl 3-aminobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, methyl ester, cbmicro_030710, 2-carbomethoxy-3-aminobenzothiophene, methyl 3-aminobenzothiophene-2-carboxylate, methyl-3-aminobenzothiophene-2-carboxylate, 3-amino-2-methoxycarbonyl-1-benzothiophene, methyl 3-amino-benzo b thiophene-2-carboxylate, methyl 3-azanyl-1-benzothiophene-2-carboxylate, amino-benzo b thiophene-2-carboxylic acid methyl ester
PubChem CID 874720
IUPAC Name methyl 3-amino-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=C(C2=CC=CC=C2S1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC432410010
SDS View Documents
Thermo Scientific Chemicals
432410010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432410050
SDS View Documents
Thermo Scientific Chemicals
432410050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

35212-85-2
207.25
VLHHEYMZLXKSQO-UHFFFAOYSA-N
874720
COC(=O)C1=C(C2=CC=CC=C2S1)N
C10H9NO2S
MFCD00206744
methyl 3-aminobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, methyl ester, cbmicro_030710, 2-carbomethoxy-3-aminobenzothiophene, methyl 3-aminobenzothiophene-2-carboxylate, methyl-3-aminobenzothiophene-2-carboxylate, 3-amino-2-methoxycarbonyl-1-benzothiophene, methyl 3-amino-benzo b thiophene-2-carboxylate, methyl 3-azanyl-1-benzothiophene-2-carboxylate, amino-benzo b thiophene-2-carboxylic acid methyl ester
methyl 3-amino-1-benzothiophene-2-carboxylate

Specifications

35212-85-2
96% min. (HPLC)
C10H9NO2S
1g
VLHHEYMZLXKSQO-UHFFFAOYSA-N
methyl 3-amino-1-benzothiophene-2-carboxylate
874720
97%
Authentic
Glass bottle
MFCD00206744
methyl 3-aminobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, methyl ester, cbmicro_030710, 2-carbomethoxy-3-aminobenzothiophene, methyl 3-aminobenzothiophene-2-carboxylate, methyl-3-aminobenzothiophene-2-carboxylate, 3-amino-2-methoxycarbonyl-1-benzothiophene, methyl 3-amino-benzo b thiophene-2-carboxylate, methyl 3-azanyl-1-benzothiophene-2-carboxylate, amino-benzo b thiophene-2-carboxylic acid methyl ester
COC(=O)C1=C(C2=CC=CC=C2S1)N
207.25
207.25
Methyl 3-aminobenzo[b]thiophene-2-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only