missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-6-bromopyrazine-2-carboxylate 98.0+%, TCI America™

Catalog No. M26555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26555G 5 g
M26551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26555G Supplier TCI America Supplier No. M26555G
Only null left

Chemical Identifiers

CAS 6966-01-4
Molecular Formula C6H6BrN3O2
Molecular Weight (g/mol) 232.04
MDL Number MFCD00834964
InChI Key CNXSIRHOIFRMOB-UHFFFAOYSA-N
Synonym methyl3-amino-6-bromopyrazine-2-carboxylate, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 2-amino-5-bromo-3-methoxycarbonyl pyrazine, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, pubchem22386
PubChem CID 227251
IUPAC Name methyl 3-amino-6-bromopyrazine-2-carboxylate
SMILES COC(=O)C1=NC(=CN=C1N)Br

Specifications

Melting Point 174°C
Quantity 5 g
Formula Weight 232.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Amino-6-bromopyrazine-2-carboxylate