Learn More
Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%
CAS: 76575-71-8 | C7H9NO2S | 171.21 g/mol
$301.86 - $1011.52
Chemical Identifiers
| CAS | 76575-71-8 |
|---|---|
| Molecular Formula | C7H9NO2S |
| Molecular Weight (g/mol) | 171.21 |
| MDL Number | MFCD00130095 |
| InChI Key | FVKMOPIFLCMZMI-UHFFFAOYSA-N |
| Synonym | 3-amino-5-methyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-methyl thiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-5-methyl-, methyl ester, methyl3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-thiophene-2-carboxylic acid, methyl 3-amino-5-methylthiophen-2-carboxylate, methyl-3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-2-thiophene-carboxylic acid, methyl ester |
| PubChem CID | 818952 |
| IUPAC Name | methyl 3-amino-5-methylthiophene-2-carboxylate |
| SMILES | COC(=O)C1=C(N)C=C(C)S1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6371903
|
Thermo Scientific Chemicals
H6371903 |
1 g |
Each for $301.86
|
|
|||||
|
AAH6371906
|
Thermo Scientific Chemicals
H6371906 |
5 g |
Each for $1,011.52
|
|
|||||
Description
Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 76575-71-8 | |
| 171.21 | |
| FVKMOPIFLCMZMI-UHFFFAOYSA-N | |
| 818952 | |
| COC(=O)C1=C(N)C=C(C)S1 |
| C7H9NO2S | |
| MFCD00130095 | |
| 3-amino-5-methyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-methyl thiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-5-methyl-, methyl ester, methyl3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-thiophene-2-carboxylic acid, methyl 3-amino-5-methylthiophen-2-carboxylate, methyl-3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-2-thiophene-carboxylic acid, methyl ester | |
| methyl 3-amino-5-methylthiophene-2-carboxylate |
Specifications
| 76575-71-8 | |
| MFCD00130095 | |
| 3-amino-5-methyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-methyl thiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-5-methyl-, methyl ester, methyl3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-thiophene-2-carboxylic acid, methyl 3-amino-5-methylthiophen-2-carboxylate, methyl-3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-2-thiophene-carboxylic acid, methyl ester | |
| FVKMOPIFLCMZMI-UHFFFAOYSA-N | |
| methyl 3-amino-5-methylthiophene-2-carboxylate | |
| 818952 | |
| 97% | |
| Methyl 3-amino-5-methylthiophene-2-carboxylate |
| C7H9NO2S | |
| 1 g | |
| Slightly soluble in water. | |
| COC(=O)C1=C(N)C=C(C)S1 | |
| 171.21 | |
| 171.22 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only