missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%

Catalog No. AAH6371906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6371906 5 g
AAH6371903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6371906 Supplier Thermo Scientific Chemicals Supplier No. H6371906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 76575-71-8 | C7H9NO2S | 171.21 g/mol

Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 76575-71-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00130095
InChI Key FVKMOPIFLCMZMI-UHFFFAOYSA-N
Synonym 3-amino-5-methyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-methyl thiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-5-methyl-, methyl ester, methyl3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-thiophene-2-carboxylic acid, methyl 3-amino-5-methylthiophen-2-carboxylate, methyl-3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-2-thiophene-carboxylic acid, methyl ester
PubChem CID 818952
IUPAC Name methyl 3-amino-5-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C=C(C)S1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 171.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-amino-5-methylthiophene-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only