missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate, 97%

Catalog No. p-7030078
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2761403 1 g
AAH2761406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2761403 Supplier Thermo Scientific Chemicals Supplier No. H2761403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1458-18-0 | C6H5Cl2N3O2 | 222.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1458-18-0
Molecular Formula C6H5Cl2N3O2
Molecular Weight (g/mol) 222.025
MDL Number MFCD00010431
InChI Key USYMCUGEGUFUBI-UHFFFAOYSA-N
Synonym methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate
PubChem CID 73828
IUPAC Name methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
SMILES COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N

Specifications

Melting Point 230°C to 233°C
Quantity 1 g
Formula Weight 222.03
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 215-947-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only