missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1458-18-0
Molecular Formula C6H5Cl2N3O2
Molecular Weight (g/mol) 222.03
MDL Number MFCD00010431
InChI Key USYMCUGEGUFUBI-UHFFFAOYSA-N
Synonym methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate
PubChem CID 73828
IUPAC Name methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
SMILES COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368280050
SDS View Documents
Thermo Scientific Chemicals
368280050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1458-18-0
222.03
USYMCUGEGUFUBI-UHFFFAOYSA-N
73828
COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N
C6H5Cl2N3O2
MFCD00010431
methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

Specifications

1458-18-0
97.5% min. (HPLC)
C6H5Cl2N3O2
5g
USYMCUGEGUFUBI-UHFFFAOYSA-N
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
73828
98%
Authentic
Glass bottle
MFCD00010431
methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate
COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N
222.03
222.03
Methyl 3-amino-5, 6-dichloro-2-pyrazinecarboxylate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environ

GHS Signal Word: Warning

EINECSNumber : 215-947-7

SDS
Documents

Documents

RUO – Research Use Only