missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (3,5-Dimethoxyphenyl)acetate 98.0+%, TCI America™
$154.71 - $462.29
Chemical Identifiers
| CAS | 6512-32-9 |
|---|---|
| Molecular Formula | C11H14O4 |
| Molecular Weight (g/mol) | 210.229 |
| MDL Number | MFCD00089971 |
| InChI Key | CLZWNNSTRIEAMH-UHFFFAOYSA-N |
| Synonym | (3,5-Dimethoxyphenyl)acetic Acid Methyl Ester |
| PubChem CID | 592191 |
| IUPAC Name | methyl 2-(3,5-dimethoxyphenyl)acetate |
| SMILES | COC1=CC(=CC(=C1)CC(=O)OC)OC |
Chemical Identifiers
| 6512-32-9 | |
| 210.229 | |
| CLZWNNSTRIEAMH-UHFFFAOYSA-N | |
| 592191 | |
| COC1=CC(=CC(=C1)CC(=O)OC)OC |
| C11H14O4 | |
| MFCD00089971 | |
| (3,5-Dimethoxyphenyl)acetic Acid Methyl Ester | |
| methyl 2-(3,5-dimethoxyphenyl)acetate |
Specifications
| 6512-32-9 | |
| C11H14O4 | |
| 1 g | |
| CLZWNNSTRIEAMH-UHFFFAOYSA-N | |
| methyl 2-(3,5-dimethoxyphenyl)acetate | |
| 592191 | |
| ≥98.0% (GC) | |
| Methyl (3,5-Dimethoxyphenyl)acetate |
| 94°C | |
| MFCD00089971 | |
| (3,5-Dimethoxyphenyl)acetic Acid Methyl Ester | |
| COC1=CC(=CC(=C1)CC(=O)OC)OC | |
| 210.229 | |
| 210.23 | |
| Liquid |
Safety and Handling
TSCA : No