missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate 98.0+%, TCI America™
$141.67 - $417.56
Chemical Identifiers
| CAS | 1458-01-1 |
|---|---|
| Molecular Formula | C6H7ClN4O2 |
| Molecular Weight (g/mol) | 202.60 |
| MDL Number | MFCD01928388 |
| InChI Key | KOOBYHRLTYIPTH-UHFFFAOYSA-N |
| Synonym | unii-4kat2q9chl, methyl 3,5-diamino-6-chloro-2-pyrazinecarboxylate, 4kat2q9chl, methyl 3, 5-diamino-6-chloropyrazine-2-carboxylate, pyrazinecarboxylic acid, 3,5-diamino-6-chloro-, methyl ester, methyl-3,5-diamino-6-chloropyrazine-2-carboxylate, 3,5-diamino-6-chloropyrazine-2-carboxylic acid methyl ester, pubchem11053, acmc-209cvs, methyl 3,5-diamino-6-chloropyrazinecarboxylate |
| PubChem CID | 73827 |
| IUPAC Name | methyl 3,5-diamino-6-chloropyrazine-2-carboxylate |
| SMILES | COC(=O)C1=NC(Cl)=C(N)N=C1N |
Chemical Identifiers
| 1458-01-1 | |
| 202.60 | |
| KOOBYHRLTYIPTH-UHFFFAOYSA-N | |
| 73827 | |
| COC(=O)C1=NC(Cl)=C(N)N=C1N |
| C6H7ClN4O2 | |
| MFCD01928388 | |
| unii-4kat2q9chl, methyl 3,5-diamino-6-chloro-2-pyrazinecarboxylate, 4kat2q9chl, methyl 3, 5-diamino-6-chloropyrazine-2-carboxylate, pyrazinecarboxylic acid, 3,5-diamino-6-chloro-, methyl ester, methyl-3,5-diamino-6-chloropyrazine-2-carboxylate, 3,5-diamino-6-chloropyrazine-2-carboxylic acid methyl ester, pubchem11053, acmc-209cvs, methyl 3,5-diamino-6-chloropyrazinecarboxylate | |
| methyl 3,5-diamino-6-chloropyrazine-2-carboxylate |
Specifications
| 1458-01-1 | |
| C6H7ClN4O2 | |
| 5 g | |
| KOOBYHRLTYIPTH-UHFFFAOYSA-N | |
| methyl 3,5-diamino-6-chloropyrazine-2-carboxylate | |
| 73827 | |
| ≥98.0% (GC,T) | |
| Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate |
| 213°C | |
| MFCD01928388 | |
| unii-4kat2q9chl, methyl 3,5-diamino-6-chloro-2-pyrazinecarboxylate, 4kat2q9chl, methyl 3, 5-diamino-6-chloropyrazine-2-carboxylate, pyrazinecarboxylic acid, 3,5-diamino-6-chloro-, methyl ester, methyl-3,5-diamino-6-chloropyrazine-2-carboxylate, 3,5-diamino-6-chloropyrazine-2-carboxylic acid methyl ester, pubchem11053, acmc-209cvs, methyl 3,5-diamino-6-chloropyrazinecarboxylate | |
| COC(=O)C1=NC(Cl)=C(N)N=C1N | |
| 202.60 | |
| 202.60 | |
| Crystalline Powder |
Safety and Handling
TSCA : No