missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,5-Bis(tert-butyldiphenylsilyloxy)benzoate (ca. 20% in Toluene, ca. 0.28mol/L), TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 25 g |
| UN Number | 1993 |
| Formula Weight | 644.96 |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 3,5-Bis(tert-butyldiphenylsilyloxy)benzoate (ca. 20% in Toluene, ca. 0.28mol/L) |
Safety and Handling
| TSCA | No |