missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-methyl-1-piperazinyl)benzoate, 97%, Thermo Scientific™

$70.45 - $284.22

Chemical Identifiers

CAS 474334-89-9
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.30
MDL Number MFCD07369792
InChI Key LPOJJZXQESTVEV-UHFFFAOYSA-N
Synonym methyl 3-4-methylpiperazin-1-yl benzoate, methyl 3-4-methyl-1-piperazinyl benzoate, 1-3-carbomethoxyphenyl-4-methylpiperazine, 1-3-carbomethoxyphenyl-4-methylpipera, methyl 3-4-methylpiperazino benzoate
PubChem CID 16217966
IUPAC Name methyl 3-(4-methylpiperazin-1-yl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5577003
View Documents
Thermo Scientific Chemicals
H5577003
1 g
Each for $70.45
Only null left
 
AAH5577006
View Documents
Thermo Scientific Chemicals
H5577006
5 g
Each for $284.22
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

474334-89-9
234.30
LPOJJZXQESTVEV-UHFFFAOYSA-N
16217966
COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1
C13H18N2O2
MFCD07369792
methyl 3-4-methylpiperazin-1-yl benzoate, methyl 3-4-methyl-1-piperazinyl benzoate, 1-3-carbomethoxyphenyl-4-methylpiperazine, 1-3-carbomethoxyphenyl-4-methylpipera, methyl 3-4-methylpiperazino benzoate
methyl 3-(4-methylpiperazin-1-yl)benzoate

Specifications

474334-89-9
1.119 g/mL
>110°C (230°F)
1.563
1 g
LPOJJZXQESTVEV-UHFFFAOYSA-N
methyl 3-(4-methylpiperazin-1-yl)benzoate
16217966
97%
Methyl 3-(4-methyl-1-piperazinyl)benzoate
Colorless to Yellow
169°C to 172°C (0.9 mmHg)
C13H18N2O2
MFCD07369792
methyl 3-4-methylpiperazin-1-yl benzoate, methyl 3-4-methyl-1-piperazinyl benzoate, 1-3-carbomethoxyphenyl-4-methylpiperazine, 1-3-carbomethoxyphenyl-4-methylpipera, methyl 3-4-methylpiperazino benzoate
COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1
234.30
234.3
Liquid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'