missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(4-Methoxycarbonylphenyl)propionate 97.0+%, TCI America™
$260.97 - $883.56
Chemical Identifiers
| CAS | 40912-11-6 |
|---|---|
| Molecular Formula | C12H14O4 |
| Molecular Weight (g/mol) | 222.24 |
| MDL Number | MFCD00059305 |
| InChI Key | MLKXCCLYRPKSBM-UHFFFAOYSA-N |
| Synonym | 3-(4-Carboxyphenyl)propionic Acid Dimethyl Ester, 3-(4-Methoxycarbonylphenyl)propionic Acid Methyl Ester |
| PubChem CID | 595759 |
| IUPAC Name | methyl 4-(3-methoxy-3-oxopropyl)benzoate |
| SMILES | COC(=O)CCC1=CC=C(C=C1)C(=O)OC |
Chemical Identifiers
| 40912-11-6 | |
| 222.24 | |
| MLKXCCLYRPKSBM-UHFFFAOYSA-N | |
| 595759 | |
| COC(=O)CCC1=CC=C(C=C1)C(=O)OC |
| C12H14O4 | |
| MFCD00059305 | |
| 3-(4-Carboxyphenyl)propionic Acid Dimethyl Ester, 3-(4-Methoxycarbonylphenyl)propionic Acid Methyl Ester | |
| methyl 4-(3-methoxy-3-oxopropyl)benzoate |
Specifications
| 40912-11-6 | |
| White | |
| MFCD00059305 | |
| 3-(4-Carboxyphenyl)propionic Acid Dimethyl Ester, 3-(4-Methoxycarbonylphenyl)propionic Acid Methyl Ester | |
| COC(=O)CCC1=CC=C(C=C1)C(=O)OC | |
| 222.24 | |
| 222.24 | |
| Crystalline Powder |
| 33°C | |
| C12H14O4 | |
| 5 g | |
| MLKXCCLYRPKSBM-UHFFFAOYSA-N | |
| methyl 4-(3-methoxy-3-oxopropyl)benzoate | |
| 595759 | |
| ≥97.0% (T) | |
| Methyl 3-(4-Methoxycarbonylphenyl)propionate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator