missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,4,5-Trimethoxybenzoate 99.0+%, TCI America™
$29.83 - $205.05
Chemical Identifiers
| CAS | 1916-07-0 |
|---|---|
| Molecular Formula | C11H14O5 |
| Molecular Weight (g/mol) | 226.228 |
| MDL Number | MFCD00008431 |
| InChI Key | KACHFMOHOPLTNX-UHFFFAOYSA-N |
| Synonym | trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8 |
| PubChem CID | 15956 |
| IUPAC Name | methyl 3,4,5-trimethoxybenzoate |
| SMILES | COC1=CC(=CC(=C1OC)OC)C(=O)OC |
Chemical Identifiers
| 1916-07-0 | |
| 226.228 | |
| KACHFMOHOPLTNX-UHFFFAOYSA-N | |
| 15956 | |
| COC1=CC(=CC(=C1OC)OC)C(=O)OC |
| C11H14O5 | |
| MFCD00008431 | |
| trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8 | |
| methyl 3,4,5-trimethoxybenzoate |
Specifications
| 1916-07-0 | |
| White | |
| C11H14O5 | |
| 25 g | |
| KACHFMOHOPLTNX-UHFFFAOYSA-N | |
| methyl 3,4,5-trimethoxybenzoate | |
| 15956 | |
| ≥99.0% (GC) | |
| Methyl 3,4,5-Trimethoxybenzoate |
| 82°C | |
| 185°C | |
| MFCD00008431 | |
| trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8 | |
| COC1=CC(=CC(=C1OC)OC)C(=O)OC | |
| 226.228 | |
| 226.23 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DI0820000
TSCA : Yes