missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trimethoxybenzoate, 98+%

CAS: 1916-07-0 | C11H14O5 | 226.228 g/mol

$40.44 - $204.78

Chemical Identifiers

CAS 1916-07-0
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00008431
InChI Key KACHFMOHOPLTNX-UHFFFAOYSA-N
Synonym trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8
PubChem CID 15956
IUPAC Name methyl 3,4,5-trimethoxybenzoate
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1193314
View Documents
Thermo Scientific Chemicals
A1193314
25 g
Each for $40.44
Only null left
 
AAA1193322
View Documents
Thermo Scientific Chemicals
A1193322
100 g
Each for $71.85
Only null left
 
AAA1193336
View Documents
Thermo Scientific Chemicals
A1193336
500 g
Each for $204.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1916-07-0
226.228
KACHFMOHOPLTNX-UHFFFAOYSA-N
15956
COC1=CC(=CC(=C1OC)OC)C(=O)OC
C11H14O5
MFCD00008431
trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8
methyl 3,4,5-trimethoxybenzoate

Specifications

1916-07-0
274°C to 275°C
MFCD00008431
2218156
KACHFMOHOPLTNX-UHFFFAOYSA-N
methyl 3,4,5-trimethoxybenzoate
15956
≥98%
81°C to 85°C
C11H14O5
25 g
trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8
COC1=CC(=CC(=C1OC)OC)C(=O)OC
226.228
226.23
Methyl 3,4,5-trimethoxybenzoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-629-3

RTECSNumber : DI0820000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only