missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trihydroxybenzoate, 98%

CAS: 99-24-1 | C8H8O5 | 184.147 g/mol

$104.86 - $300.10

Chemical Identifiers

CAS 99-24-1
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.147
MDL Number MFCD00002194
InChI Key FBSFWRHWHYMIOG-UHFFFAOYSA-N
Synonym methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
PubChem CID 7428
IUPAC Name methyl 3,4,5-trihydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0592618
View Documents
Thermo Scientific Chemicals
L0592618
50 g
Each for $104.86
Only null left
 
AAL0592630
View Documents
Thermo Scientific Chemicals
L0592630
250 g
Each for $300.10
Only null left
 
Description

Description

Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

99-24-1
184.147
FBSFWRHWHYMIOG-UHFFFAOYSA-N
7428
COC(=O)C1=CC(=C(C(=C1)O)O)O
C8H8O5
MFCD00002194
methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
methyl 3,4,5-trihydroxybenzoate

Specifications

99-24-1
C8H8O5
50 g
methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
FBSFWRHWHYMIOG-UHFFFAOYSA-N
methyl 3,4,5-trihydroxybenzoate
7428
0.98
198°C to 203°C
MFCD00002194
144345
Slightly soluble in water.
COC(=O)C1=CC(=C(C(=C1)O)O)O
184.147
184.15
Methyl 3,4,5-trihydroxybenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H319

EINECSNumber : 202-741-7

RTECSNumber : LW8000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only