missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trihydroxybenzoate, 98%

Catalog No. AAL0592618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0592618 50 g
AAL0592630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0592618 Supplier Thermo Scientific Chemicals Supplier No. L0592618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-24-1 | C8H8O5 | 184.147 g/mol

Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 99-24-1
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.147
MDL Number MFCD00002194
InChI Key FBSFWRHWHYMIOG-UHFFFAOYSA-N
Synonym methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
PubChem CID 7428
IUPAC Name methyl 3,4,5-trihydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 198°C to 203°C
Quantity 50 g
Merck Index 144345
Solubility Information Slightly soluble in water.
Formula Weight 184.15
Percent Purity 0.98
Chemical Name or Material Methyl 3,4,5-trihydroxybenzoate
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 202-741-7
RTECSNumber LW8000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only