missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,3-Dimethoxypropionate 98.0+%, TCI America™
$202.15 - $202.15
Chemical Identifiers
| CAS | 7424-91-1 |
|---|---|
| Molecular Formula | C6H12O4 |
| Molecular Weight (g/mol) | 148.158 |
| MDL Number | MFCD00010650 |
| InChI Key | SMCVPMKCDDNUCQ-UHFFFAOYSA-N |
| PubChem CID | 81924 |
| IUPAC Name | methyl 3,3-dimethoxypropanoate |
| SMILES | COC(CC(=O)OC)OC |
Chemical Identifiers
| 7424-91-1 | |
| 148.158 | |
| SMCVPMKCDDNUCQ-UHFFFAOYSA-N | |
| methyl 3,3-dimethoxypropanoate |
| C6H12O4 | |
| MFCD00010650 | |
| 81924 | |
| COC(CC(=O)OC)OC |
Specifications
| 7424-91-1 | |
| 80°C | |
| MFCD00010650 | |
| methyl 3,3-dimethoxypropionate, 3,3-dimethoxypropionic acid methyl ester, methyl3,3-dimethoxypropionate, propanoic acid, 3,3-dimethoxy-, methyl ester, zlchem 282, pubchem20228, acmc-1bdjk, ksc490c4j, methyl 3,3-dimethoxy-propanoate, smcvpmkcddnucq-uhfffaoysa | |
| COC(CC(=O)OC)OC | |
| 148.158 | |
| 148.16 | |
| Liquid |
| Colorless | |
| C6H12O4 | |
| 25 g | |
| SMCVPMKCDDNUCQ-UHFFFAOYSA-N | |
| methyl 3,3-dimethoxypropanoate | |
| 81924 | |
| ≥98.0% (GC) | |
| Methyl 3,3-Dimethoxypropionate |
Safety and Handling
TSCA : No