missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-dimethoxypropionate, 96%

CAS: 7424-91-1 | C6H12O4 | 148.158 g/mol

$79.27 - $257.74

Chemical Identifiers

CAS 7424-91-1
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.158
MDL Number MFCD00010650
InChI Key SMCVPMKCDDNUCQ-UHFFFAOYSA-N
PubChem CID 81924
IUPAC Name methyl 3,3-dimethoxypropanoate
SMILES COC(CC(=O)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2420009
View Documents
Thermo Scientific Chemicals
B2420009
10 g
Each for $79.27
Only null left
 
AAB2420018
View Documents
Thermo Scientific Chemicals
B2420018
50 g
Each for $257.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7424-91-1
148.158
SMCVPMKCDDNUCQ-UHFFFAOYSA-N
methyl 3,3-dimethoxypropanoate
C6H12O4
MFCD00010650
81924
COC(CC(=O)OC)OC

Specifications

7424-91-1
76°C to 77°C (20 mmHg)
C6H12O4
(CH3O)2CHCH2CO2CH3
10 g
methyl 3,3-dimethoxypropionate, 3,3-dimethoxypropionic acid methyl ester, methyl3,3-dimethoxypropionate, propanoic acid, 3,3-dimethoxy-, methyl ester, zlchem 282, pubchem20228, acmc-1bdjk, ksc490c4j, methyl 3,3-dimethoxy-propanoate, smcvpmkcddnucq-uhfffaoysa
COC(CC(=O)OC)OC
148.158
148.16
Methyl 3,3-dimethoxypropionate
1.06
65°C (149°F)
1.41
MFCD00010650
1561517
SMCVPMKCDDNUCQ-UHFFFAOYSA-N
methyl 3,3-dimethoxypropanoate
81924
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 231-055-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only