missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-dimethoxypropionate, 96%

Catalog No. AAB2420009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2420009 10 g
AAB2420018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2420009 Supplier Thermo Scientific Chemicals Supplier No. B2420009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7424-91-1 | C6H12O4 | 148.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7424-91-1
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.158
MDL Number MFCD00010650
InChI Key SMCVPMKCDDNUCQ-UHFFFAOYSA-N
PubChem CID 81924
IUPAC Name methyl 3,3-dimethoxypropanoate
SMILES COC(CC(=O)OC)OC

Specifications

Density 1.06
Boiling Point 76°C to 77°C (20 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.41
Linear Formula (CH3O)2CHCH2CO2CH3
Quantity 10 g
Beilstein 1561517
Synonym methyl 3,3-dimethoxypropionate, 3,3-dimethoxypropionic acid methyl ester, methyl3,3-dimethoxypropionate, propanoic acid, 3,3-dimethoxy-, methyl ester, zlchem 282, pubchem20228, acmc-1bdjk, ksc490c4j, methyl 3,3-dimethoxy-propanoate, smcvpmkcddnucq-uhfffaoysa
Formula Weight 148.16
Percent Purity 96%
Chemical Name or Material Methyl 3,3-dimethoxypropionate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 231-055-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only