missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,3,3-Trifluoropropionate 98.0+%, TCI America™
$175.22 - $581.59
Chemical Identifiers
| CAS | 18830-44-9 |
|---|---|
| Molecular Formula | C4H5F3O2 |
| Molecular Weight (g/mol) | 142.077 |
| MDL Number | MFCD02262206 |
| InChI Key | PMGBATZKLCISOD-UHFFFAOYSA-N |
| Synonym | 3,3,3-Trifluoropropionic Acid Methyl Ester |
| PubChem CID | 2782392 |
| IUPAC Name | methyl 3,3,3-trifluoropropanoate |
| SMILES | COC(=O)CC(F)(F)F |
Chemical Identifiers
| 18830-44-9 | |
| 142.077 | |
| PMGBATZKLCISOD-UHFFFAOYSA-N | |
| 2782392 | |
| COC(=O)CC(F)(F)F |
| C4H5F3O2 | |
| MFCD02262206 | |
| 3,3,3-Trifluoropropionic Acid Methyl Ester | |
| methyl 3,3,3-trifluoropropanoate |
Specifications
| 18830-44-9 | |
| 96°C | |
| MFCD02262206 | |
| 3272 | |
| PMGBATZKLCISOD-UHFFFAOYSA-N | |
| methyl 3,3,3-trifluoropropanoate | |
| 2782392 | |
| ≥98.0% (GC) | |
| Methyl 3,3,3-Trifluoropropionate |
| Colorless | |
| C4H5F3O2 | |
| 1 g | |
| 3,3,3-Trifluoropropionic Acid Methyl Ester | |
| COC(=O)CC(F)(F)F | |
| 142.077 | |
| 142.08 | |
| Liquid |
Safety and Handling
TSCA : No