Learn More
Methyl 2-pyridyl ketoxime, 97%
CAS: 1758-54-9 | C7H8N2O | 136.15 g/mol
$91.95 - $307.68
Chemical Identifiers
| CAS | 1758-54-9 |
|---|---|
| Molecular Formula | C7H8N2O |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00605540 |
| InChI Key | KWYHBFKEFFMBHW-SREVYHEPSA-N |
| Synonym | 1-pyridin-2-yl ethanone oxime, 1-2-pyridinyl-1-ethanone oxime, 1-2-pyridinyl ethanone oxime, 1-pyridin-2-yl-ethanone oxime, 1-pyridin-2-ylethan-1-one oxime, e-1-pyridin-2-yl ethanone oxime, e-n-1-pyridin-2-yl ethylidene hydroxylamine, methyl2-pyridylketoneoxime, 2-1-nitrosoethylidene-1h-pyridine, e-1-pyridine-2-yl ethanone oxime |
| PubChem CID | 6740780 |
| SMILES | C\C(N=O)=C1\NC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5223203
|
Thermo Scientific Chemicals
H5223203 |
1 g |
Each for $91.95
|
|
|||||
|
AAH5223206
|
Thermo Scientific Chemicals
H5223206 |
5 g |
Each for $307.68
|
|
|||||
Description
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 1758-54-9 | |
| 136.15 | |
| KWYHBFKEFFMBHW-SREVYHEPSA-N | |
| 6740780 |
| C7H8N2O | |
| MFCD00605540 | |
| 1-pyridin-2-yl ethanone oxime, 1-2-pyridinyl-1-ethanone oxime, 1-2-pyridinyl ethanone oxime, 1-pyridin-2-yl-ethanone oxime, 1-pyridin-2-ylethan-1-one oxime, e-1-pyridin-2-yl ethanone oxime, e-n-1-pyridin-2-yl ethylidene hydroxylamine, methyl2-pyridylketoneoxime, 2-1-nitrosoethylidene-1h-pyridine, e-1-pyridine-2-yl ethanone oxime | |
| C\C(N=O)=C1\NC=CC=C1 |
Specifications
| 1758-54-9 | |
| MFCD00605540 | |
| 1-pyridin-2-yl ethanone oxime, 1-2-pyridinyl-1-ethanone oxime, 1-2-pyridinyl ethanone oxime, 1-pyridin-2-yl-ethanone oxime, 1-pyridin-2-ylethan-1-one oxime, e-1-pyridin-2-yl ethanone oxime, e-n-1-pyridin-2-yl ethylidene hydroxylamine, methyl2-pyridylketoneoxime, 2-1-nitrosoethylidene-1h-pyridine, e-1-pyridine-2-yl ethanone oxime | |
| KWYHBFKEFFMBHW-SREVYHEPSA-N | |
| (2Z)-2-(1-nitrosoethylidene)-1,2-dihydropyridine | |
| 6740780 | |
| 97% |
| C7H8N2O | |
| 1 g | |
| Slightly soluble in water. | |
| C\C(N=O)=C1\NC=CC=C1 | |
| 136.15 | |
| 136.15 | |
| Methyl 2-pyridyl ketoxime |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only