missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-pyridyl ketoxime, 97%

Catalog No. AAH5223206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5223206 5 g
AAH5223203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5223206 Supplier Thermo Scientific Chemicals Supplier No. H5223206

CAS: 1758-54-9 | C7H8N2O | 136.15 g/mol

Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1758-54-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00605540
InChI Key KWYHBFKEFFMBHW-SREVYHEPSA-N
Synonym 1-pyridin-2-yl ethanone oxime, 1-2-pyridinyl-1-ethanone oxime, 1-2-pyridinyl ethanone oxime, 1-pyridin-2-yl-ethanone oxime, 1-pyridin-2-ylethan-1-one oxime, e-1-pyridin-2-yl ethanone oxime, e-n-1-pyridin-2-yl ethylidene hydroxylamine, methyl2-pyridylketoneoxime, 2-1-nitrosoethylidene-1h-pyridine, e-1-pyridine-2-yl ethanone oxime
PubChem CID 6740780
SMILES C\C(N=O)=C1\NC=CC=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
IUPAC Name 2-(1-nitrosoethylidene)-1H-pyridine
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material Methyl 2-pyridyl ketoxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only