missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2-pentanone, 98.5% min., ACS, MilliporeSigma™

$13159.79 - $13159.79

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MMX130030
View Documents
Technidata
MX130030
207 L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

108-10-1
100.161
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CHEBI:82344
CC(C)CC(=O)C
C6H12O
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one

Specifications

108-10-1
C6H12O
207 L
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
1.16
Liquid
98.5%, min.
(CH3)2CHCH2COCH3
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CC(C)CC(=O)C
100.161
CHEBI:82344
ACS/NF/USP
Methyl-2-pentanone, For ACS analysis
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents