missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2-pentanone, 98.5% min., ACS, MilliporeSigma™

Catalog No. MMX130030
Change view
Click to view available options
Quantity:
207 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX130030 207 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX130030 Supplier Technidata Supplier No. MX130030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Assay Percent Range 98.5%, min.
Linear Formula (CH3)2CHCH2COCH3
Quantity 207 L
Formula Weight 1.16
Grade ACS/NF/USP
Physical Form Liquid
Chemical Name or Material Methyl-2-pentanone, For ACS analysis
Recommended Storage Room Temperature