missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(p-Tolyl)benzoate 98.0+%, TCI America™
$70.84 - $283.33
Chemical Identifiers
| CAS | 114772-34-8 |
|---|---|
| Molecular Formula | C15H14O2 |
| Molecular Weight (g/mol) | 226.28 |
| MDL Number | MFCD03453661 |
| InChI Key | IHNIAWHITVGYJJ-UHFFFAOYSA-N |
| PubChem CID | 1393873 |
| IUPAC Name | methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate |
| SMILES | COC(=O)C1=CC=CC=C1C1=CC=C(C)C=C1 |
Chemical Identifiers
| 114772-34-8 | |
| 226.28 | |
| IHNIAWHITVGYJJ-UHFFFAOYSA-N | |
| methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate |
| C15H14O2 | |
| MFCD03453661 | |
| 1393873 | |
| COC(=O)C1=CC=CC=C1C1=CC=C(C)C=C1 |
Specifications
| 114772-34-8 | |
| White-Yellow | |
| MFCD03453661 | |
| 2-(p-Tolyl)benzoic Acid Methyl Ester, Methyl 2-(4-Methylphenyl)benzoate, Methyl 4′C-Methylbiphenyl-2-carboxylate, 4′C-Methylbiphenyl-2-carboxylic Acid Methyl Ester | |
| COC(=O)C1=CC=CC=C1C1=CC=C(C)C=C1 | |
| 226.28 | |
| 226.28 | |
| Crystalline Powder |
| 59°C | |
| C15H14O2 | |
| 5 g | |
| IHNIAWHITVGYJJ-UHFFFAOYSA-N | |
| methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate | |
| 1393873 | |
| ≥98.0% (GC) | |
| Methyl 2-(p-Tolyl)benzoate |
Safety and Handling
TSCA : No