missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(p-Tolyl)benzoate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 59°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | 2-(p-Tolyl)benzoic Acid Methyl Ester, Methyl 2-(4-Methylphenyl)benzoate, Methyl 4′C-Methylbiphenyl-2-carboxylate, 4′C-Methylbiphenyl-2-carboxylic Acid Methyl Ester |
| Formula Weight | 226.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 2-(p-Tolyl)benzoate |
Safety and Handling
| TSCA | No |