missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxoindoline-7-carboxylate, 96%, Thermo Scientific Chemicals

Catalog No. p-7056895
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66091MD 250 mg
AAH6609103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66091MD Supplier Thermo Scientific Chemicals Supplier No. H66091MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380427-39-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD02179610
InChI Key XVJRNLIMSQFUAI-UHFFFAOYSA-N
Synonym methyl 2-oxoindoline-7-carboxylate, methyl oxindole-7-carboxylate, methyl 2-oxo-2,3-dihydro-1h-indole-7-carboxylate, 1h-indole-7-carboxylic acid, 2,3-dihydro-2-oxo-, methyl ester, acmc-1aje4, methyl oxindole-7-carboxylate,, methyl 2-oxindole-7-carboxylate, 2-oxyindole-7-carboxylic acid methyl ester, 1h-indole-7-carboxylicacid, 2,3-dihydro-2-oxo-, methyl ester
PubChem CID 2773518
IUPAC Name methyl 2-oxo-1,3-dihydroindole-7-carboxylate
SMILES COC(=O)C1=C2C(=CC=C1)CC(=O)N2

Specifications

Quantity 250 mg
Beilstein 15864880
Formula Weight 191.19
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 2-oxoindoline-7-carboxylate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only