missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Oxobutyrate 97.0+%, TCI America™

Catalog No. M236925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M236925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M236925G Supplier TCI America Supplier No. M236925G
Only null left

Chemical Identifiers

CAS 3952-66-7
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD03093092
InChI Key XPIWVCAMONZQCP-UHFFFAOYSA-N
Synonym 2-Oxobutyric Acid Methyl Ester
PubChem CID 545127
IUPAC Name methyl 2-oxobutanoate
SMILES CCC(=O)C(=O)OC

Specifications

Melting Point -47°C
Color Yellow
Boiling Point 155°C
Quantity 25 g
UN Number 1993
Formula Weight 116.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Oxobutyrate
EINECSNumber (2)-3952
TSCA No