missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxo-2H-pyran-3-carboxylate, 98%

Catalog No. p-7028550
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2593203 1 g
AAH2593206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2593203 Supplier Thermo Scientific Chemicals Supplier No. H2593203

CAS: 25991-27-9 | C7H6O4 | 154.121 g/mol

Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 25991-27-9
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00075443
InChI Key GJVZWOMUTYNUCE-UHFFFAOYSA-N
Synonym methyl 2-oxo-2h-pyran-3-carboxylate, 3-carbomethoxy-2-pyrone, methyl 2-pyrone-3-carboxylate, 2-oxo-2h-pyran-3-carboxylic acid methyl ester, 2h-pyran-3-carboxylic acid, 2-oxo-, methyl ester, methoxycarbonyl pyranone, acmc-1clg5, ksc496k6b
PubChem CID 117704
IUPAC Name methyl 2-oxopyran-3-carboxylate
SMILES COC(=O)C1=CC=COC1=O

Specifications

Melting Point 74°C to 77°C
Boiling Point 146°C to 148°C (0.75 mmHg)
Quantity 1 g
Beilstein 1424749
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material Methyl 2-oxo-2H-pyran-3-carboxylate
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 247-397-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only