missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Octynoate 98.0+%, TCI America™

Catalog No. O023925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O023925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O023925ML Supplier TCI America Supplier No. O023925ML

Chemical Identifiers

CAS 111-12-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00009530
InChI Key FRLZQXRXIKQFNS-UHFFFAOYSA-N
Synonym methyl 2-octynoate, folione, methyl heptine carbonate, methyl 2-octynate, methyl 2-octinate, 2-octynoic acid, methyl ester, vert de violette, artificial, methyl hept-1-yne-1-carboxylate, methyl pentylacetylenecarboxylate, unii-0ttp6yt2t3
PubChem CID 8092
IUPAC Name methyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OC

Specifications

Color Colorless
Boiling Point 220°C
Quantity 25 mL
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Octynoate
EINECSNumber (2)-2610
RTECSNumber RI2735000
TSCA Yes