missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-octynoate, 98%

Catalog No. p-7025639
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AAB2124214 25 g
AAB2124222 100 g
2 options

Catalog No. AAB2124214

Supplier: Thermo Scientific Chemicals B2124214

Only null left

CAS: 111-12-6 | C9H14O2 | 154.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-12-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00009530
InChI Key FRLZQXRXIKQFNS-UHFFFAOYSA-N
Synonym methyl 2-octynoate, folione, methyl heptine carbonate, methyl 2-octynate, methyl 2-octinate, 2-octynoic acid, methyl ester, vert de violette, artificial, methyl hept-1-yne-1-carboxylate, methyl pentylacetylenecarboxylate, unii-0ttp6yt2t3
PubChem CID 8092
IUPAC Name methyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OC

Specifications

Density 0.92
Boiling Point 216°C to 218°C
Flash Point 88°C (192°F)
Refractive Index 1.446
Quantity 25 g
Beilstein 1756887
Formula Weight 154.2
Percent Purity 98%
Chemical Name or Material Methyl 2-octynoate
Hazard Category H227-H317
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
EINECSNumber 203-836-6
RTECSNumber RI2735000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only