missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Nonynoate 98.0+%, TCI America™

Catalog No. N059525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N059525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N059525ML Supplier TCI America Supplier No. N059525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-80-8
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00009547
InChI Key NTLJTUMJJWVCTL-UHFFFAOYSA-N
Synonym methyl 2-nonynoate, 2-nonynoic acid methyl ester, methyl octine carbonate, methyl octyne carbonate, 2-nonynoic acid, methyl ester, methyloctyne carboxylate, methyloctyne carbonate, methyl octin carbonate, octynecarboxylic acid, methyl ester, methyl octynecarboxylate
PubChem CID 8137
ChEBI CHEBI:51749
IUPAC Name methyl non-2-ynoate
SMILES CCCCCCC#CC(=O)OC

Specifications

Color Yellow
Boiling Point 121°C
Quantity 25 mL
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Nonynoate
EINECSNumber (2)-1073&(2)-2609&(2)-2611
RTECSNumber RB3450000
TSCA Yes