missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nonynoate, 98%, Thermo Scientific Chemicals

Catalog No. AAB2140914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2140914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2140914 Supplier Thermo Scientific Chemicals Supplier No. B2140914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-80-8 | Molecular Formula: C10H16O2 | Molecular Weight: 168.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-80-8
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00009547
InChI Key NTLJTUMJJWVCTL-UHFFFAOYSA-N
Synonym methyl 2-nonynoate, 2-nonynoic acid methyl ester, methyl octine carbonate, methyl octyne carbonate, 2-nonynoic acid, methyl ester, methyloctyne carboxylate, methyloctyne carbonate, methyl octin carbonate, octynecarboxylic acid, methyl ester, methyl octynecarboxylate
PubChem CID 8137
ChEBI CHEBI:51749
IUPAC Name methyl non-2-ynoate
SMILES CCCCCCC#CC(=O)OC

Specifications

Density 0.915
Boiling Point 121°C (20 mmHg)
Flash Point 104°C (219°F)
Odor Floral
Refractive Index 1.4485
Linear Formula CH3(CH2)5C≡CCO2CH3
Quantity 25 g
Beilstein 1759870
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Methyl 2-nonynoate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 203-909-2
RTECSNumber RB3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only