Learn More
Methyl 2-methylbutyrate, 98%
CAS: 868-57-5 | C6H12O2 | 116.16 g/mol
$123.04 - $123.04
Chemical Identifiers
| CAS | 868-57-5 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00009335 |
| InChI Key | OCWLYWIFNDCWRZ-UHFFFAOYNA-N |
| Synonym | methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate |
| PubChem CID | 13357 |
| IUPAC Name | methyl 2-methylbutanoate |
| SMILES | CCC(C)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0810322
|
Thermo Scientific Chemicals
L0810322 |
100 g |
Each for $123.04
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 868-57-5 | |
| 116.16 | |
| OCWLYWIFNDCWRZ-UHFFFAOYNA-N | |
| 13357 | |
| CCC(C)C(=O)OC |
| C6H12O2 | |
| MFCD00009335 | |
| methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate | |
| methyl 2-methylbutanoate |
Specifications
| 868-57-5 | |
| 114°C to 116°C | |
| Fruit-like | |
| 1.394 | |
| 100 g | |
| 1720409 | |
| OCWLYWIFNDCWRZ-UHFFFAOYNA-N | |
| methyl 2-methylbutanoate | |
| 13357 | |
| 98% |
| 0.885 | |
| 16°C (61°F) | |
| C6H12O2 | |
| MFCD00009335 | |
| UN3272 | |
| methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate | |
| CCC(C)C(=O)OC | |
| 116.16 | |
| 116.16 | |
| Methyl 2-methylbutyrate |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H225-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 212-778-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only