missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methyl-3-furoate, 97%

Catalog No. p-7081794
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1197914 25 g
AAA1197922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1197914 Supplier Thermo Scientific Chemicals Supplier No. A1197914
Only null left

CAS: 6141-58-8 | C7H8O3 | 140.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6141-58-8
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003249
InChI Key UVRRIABXNIGUJZ-UHFFFAOYSA-N
PubChem CID 80237
IUPAC Name methyl 2-methylfuran-3-carboxylate
SMILES CC1=C(C=CO1)C(=O)OC

Specifications

Density 1.114
Boiling Point 71°C to 72°C (20 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.4745
Quantity 25 g
Beilstein 1342040
Synonym methyl 2-methyl-3-furoate, methyl 2-methyl-3-furancarboxylate, 2-methyl-furan-3-carboxylic acid methyl ester, 3-furancarboxylic acid, 2-methyl-, methyl ester, 3-furoic acid, 2-methyl-, methyl ester, methyl2-methyl-3-furoate, acmc-1bcvo, methyl 2-methyl-3-furoate #, methyl-2-methyl-3-furoate, 2-methylfuran-3-carboxylic acid methyl
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Methyl 2-methyl-3-furoate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 228-132-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only