missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxynicotinate, 97%

Catalog No. AAH3333403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3333403 1 g
AAH3333406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3333403 Supplier Thermo Scientific Chemicals Supplier No. H3333403
Only null left

CAS: 10128-91-3 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10128-91-3
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00661282
InChI Key SILBTMNCGYLTOK-UHFFFAOYSA-N
Synonym methyl 2-hydroxynicotinate, methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate, methyl 2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 2-hydroxypyridine-3-carboxylate, 2-hydroxy-nicotinic acid methyl ester, 2-hydroxynicotinic acid methyl ester, 2-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 2-hydroxy-, methyl ester, methyl 2-hydroxy-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-, methyl ester
PubChem CID 579028
IUPAC Name methyl 2-oxo-1H-pyridine-3-carboxylate
SMILES COC(=O)C1=CC=CNC1=O

Specifications

Melting Point 153°C to 155°C
Color Cream
Quantity 1 g
Formula Weight 153.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-hydroxynicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only