missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Hydroxyisobutyrate 99.5+%, TCI America™
$74.56 - $596.49
Chemical Identifiers
| CAS | 2110-78-3 |
|---|---|
| Molecular Formula | C5H10O3 |
| Molecular Weight (g/mol) | 118.132 |
| MDL Number | MFCD00004457 |
| InChI Key | XYVQFUJDGOBPQI-UHFFFAOYSA-N |
| PubChem CID | 75024 |
| IUPAC Name | methyl 2-hydroxy-2-methylpropanoate |
| SMILES | CC(C)(C(=O)OC)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H025625ML
|
TCI America
H025625ML |
25 mL |
Each for $74.56
|
|
|||||
|
H0256100ML
|
TCI America
H0256100ML |
100 mL |
Each for $206.90
|
|
|||||
|
H0256500ML
|
TCI America
H0256500ML |
500 mL |
Each for $596.49
|
|
|||||
Chemical Identifiers
| 2110-78-3 | |
| 118.132 | |
| XYVQFUJDGOBPQI-UHFFFAOYSA-N | |
| methyl 2-hydroxy-2-methylpropanoate |
| C5H10O3 | |
| MFCD00004457 | |
| 75024 | |
| CC(C)(C(=O)OC)O |
Specifications
| 2110-78-3 | |
| 137°C | |
| MFCD00004457 | |
| 1993 | |
| XYVQFUJDGOBPQI-UHFFFAOYSA-N | |
| methyl 2-hydroxy-2-methylpropanoate | |
| 75024 | |
| ≥99.5% (GC) | |
| Methyl 2-Hydroxyisobutyrate |
| Colorless | |
| C5H10O3 | |
| 25 mL | |
| methyl 2-hydroxyisobutyrate, methyl 2-methyllactate, 2-hydroxyisobutyric acid methyl ester, methyl 2-hydroxy-2-methylpropionate, methyl2-hydroxyisobutyrate, propanoic acid, 2-hydroxy-2-methyl-, methyl ester, methyl alpha-hydroxyisobutyrate, methyl .alpha.-hydroxyisobutyrate, methyl ester of 2-methyllactic acid, methyl 2-hydroxy-2-methyl-propanoate | |
| CC(C)(C(=O)OC)O | |
| 118.132 | |
| 118.13 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1404&(2)-1430
TSCA : Yes