missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Hydroxyisobutyrate 99.5+%, TCI America™

Catalog No. H0256500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0256500ML 500 mL
H025625ML 25 mL
H0256100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0256500ML Supplier TCI America Supplier No. H0256500ML

Chemical Identifiers

CAS 2110-78-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00004457
InChI Key XYVQFUJDGOBPQI-UHFFFAOYSA-N
PubChem CID 75024
IUPAC Name methyl 2-hydroxy-2-methylpropanoate
SMILES CC(C)(C(=O)OC)O

Specifications

Color Colorless
Boiling Point 137°C
Quantity 500 mL
UN Number 1993
Synonym methyl 2-hydroxyisobutyrate, methyl 2-methyllactate, 2-hydroxyisobutyric acid methyl ester, methyl 2-hydroxy-2-methylpropionate, methyl2-hydroxyisobutyrate, propanoic acid, 2-hydroxy-2-methyl-, methyl ester, methyl alpha-hydroxyisobutyrate, methyl .alpha.-hydroxyisobutyrate, methyl ester of 2-methyllactic acid, methyl 2-hydroxy-2-methyl-propanoate
Formula Weight 118.13
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Hydroxyisobutyrate
EINECSNumber (2)-1404&(2)-1430
TSCA Yes